Assay Details
| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177158.5 | NP_796132.2 | 9 - 10 | 1416 | 88 | |
| GenBank mRNA | AK030993.1 | - | 9 - 10 | 1409 | 88 | |
| AK082828.1 | - | 2 - 3 | 652 | 88 | ||
| AK162469.1 | - | 2 - 3 | 321 | 88 | ||
| AK168472.1 | - | 2 - 3 | 579 | 88 | ||
| BC044761.1 | - | 9 - 10 | 1452 | 88 | ||
| BC046530.1 | - | 9 - 10 | 1452 | 5329601 | 88 | |
More Information
Set Membership: |
|

Rinl
Genomic Map