Assay Details
| Gene Symbol: Sars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023637.3 | NP_076126.2 | 5 - 6 | 641 | 74 | |
| GenBank mRNA | AB029949.1 | - | 5 - 6 | 613 | 74 | |
| AK010491.1 | - | 5 - 6 | 619 | 74 | ||
| AK087606.1 | - | 5 - 6 | 608 | 74 | ||
| AK140760.1 | - | 5 - 6 | 608 | 74 | ||
| BC079664.1 | - | 5 - 6 | 618 | 30357834 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sars2
Genomic Map