Assay Details
| Gene Symbol: Mtnr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145712.2 | NP_663758.2 | 1 - 2 | 317 | 58 | |
| GenBank mRNA | AB377276.1 | - | 317 | 58 | ||
| AY145850.1 | - | 1 - 2 | 221 | 58 | ||
| BC104324.1 | - | 1 - 2 | 221 | 40044921 | 58 | |
| BC104325.1 | - | 1 - 2 | 221 | 40044928 | 58 | |
More Information
Set Membership: |
|

Mtnr1b
Genomic Map