Assay Details
| Gene Symbol: Paf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019458.3 | NP_062331.2 | 6 - 7 | 675 | 115 | |
| GenBank mRNA | AB041615.1 | - | 6 - 7 | 693 | 115 | |
| AK017762.1 | - | 6 - 7 | 745 | 115 | ||
| AK134857.1 | - | 6 - 7 | 688 | 115 | ||
| AK148538.1 | - | 6 - 7 | 710 | 115 | ||
| AK151665.1 | - | 6 - 7 | 721 | 115 | ||
| AK152778.1 | - | 6 - 7 | 691 | 115 | ||
| AK152836.1 | - | 6 - 7 | 721 | 115 | ||
| BC010317.1 | - | 5 - 6 | 719 | 115 | ||
| BC029843.1 | - | 6 - 7 | 675 | 2651081 | 115 | |
| BC083337.1 | - | 6 - 7 | 656 | 5701517 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Paf1
Genomic Map