Assay Details
| Gene Symbol: Paf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019458.3 | NP_062331.2 | 9 - 10 | 950 | 71 | |
| GenBank mRNA | AB041615.1 | - | 9 - 10 | 968 | 71 | |
| AK017762.1 | - | 9 - 10 | 1020 | 71 | ||
| AK134857.1 | - | 9 - 10 | 963 | 71 | ||
| AK148538.1 | - | 9 - 10 | 985 | 71 | ||
| AK151665.1 | - | 9 - 10 | 996 | 71 | ||
| AK152778.1 | - | 9 - 10 | 966 | 71 | ||
| AK152836.1 | - | 9 - 10 | 996 | 71 | ||
| BC010317.1 | - | 8 - 9 | 994 | 71 | ||
| BC029843.1 | - | 9 - 10 | 950 | 2651081 | 71 | |
| BC083337.1 | - | 9 - 10 | 931 | 5701517 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Paf1
Genomic Map