Assay Details
| Gene Symbol: Actn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021895.2 | NP_068695.1 | 9 - 10 | 1007 | 64 | |
| GenBank mRNA | AJ289242.2 | - | 9 - 10 | 1007 | 64 | |
| AK030237.1 | - | 2 - 3 | 177 | 64 | ||
| AK145322.1 | - | 9 - 10 | 1060 | 64 | ||
| AK150044.1 | - | 9 - 10 | 1009 | 64 | ||
| AK201509.1 | - | 1 - 2 | 70 | 64 | ||
| AK202682.1 | - | 1 - 2 | 70 | 64 | ||
| BC013616.1 | - | 9 - 10 | 1016 | 3157024 | 64 | |
| BC027032.1 | - | 9 - 10 | 956 | 64 | ||
| BC087554.1 | - | 9 - 10 | 960 | 5351470 | 64 | |
| DQ303411.1 | - | 9 - 10 | 910 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actn4
Genomic Map