Assay Details
| Gene Symbol: Unc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029770.2 | NP_084046.2 | 13 - 14 | 2588 | 95 | |
| GenBank mRNA | AJ487853.1 | - | 13 - 14 | 2491 | 95 | |
| AK018177.1 | - | 13 - 14 | 2588 | 95 | ||
| AK147603.1 | - | 12 - 13 | 2450 | 95 | ||
| AK154271.1 | - | 13 - 14 | 2588 | 95 | ||
| AK154287.1 | - | 13 - 14 | 2592 | 95 | ||
| AK158284.1 | - | 13 - 14 | 2588 | 95 | ||
| BC048162.1 | - | 12 - 13 | 2362 | 95 | ||
| BC057560.1 | - | 12 - 13 | 2362 | 6417563 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc5b
Genomic Map