Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900018102.mm.1 |
NM_145985.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018102.mm.1 |
NM_145985.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018102.mm.1 |
TC0900002175.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018104.mm.1 |
NM_145985.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018104.mm.1 |
NM_145985.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018104.mm.1 |
TC0900002175.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018105.mm.1 |
NM_145985.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018105.mm.1 |
NM_145985.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018105.mm.1 |
TC0900002175.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018103.mm.1 |
NM_145985.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018103.mm.1 |
NM_145985.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018103.mm.1 |
TC0900002175.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018101.mm.1 |
NM_145985.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018101.mm.1 |
NM_145985.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018101.mm.1 |
TC0900002175.mm.1 |
MTA TC |