Assay Details
| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143776.1 | NP_001137248.1 | 1 - 2 | 167 | 95 | |
| NM_024244.4 | NP_077206.3 | 1 - 2 | 167 | 95 | ||
| GenBank mRNA | AK004797.1 | - | 1 - 2 | 151 | 95 | |
| AK041147.1 | - | 1 - 2 | 168 | 95 | ||
| AK082202.1 | - | 1 - 2 | 163 | 95 | ||
| BB637603.1 | - | 1 - 2 | 168 | 95 | ||
| BC129839.1 | - | 1 - 2 | 123 | 40060810 | 95 | |
| BC129840.1 | - | 1 - 2 | 123 | 40060811 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam13c
Genomic Map