Assay Details
| Gene Symbol: Bmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007555.3 | NP_031581.2 | 1 - 2 | 1224 | 90 | |
| GenBank mRNA | AK033362.1 | - | 1 - 2 | 1227 | 90 | |
| AK160412.1 | - | 332 | 90 | |||
| BC100751.1 | - | 1 - 2 | 489 | 90 | ||
| BC100752.1 | - | 1 - 2 | 489 | 90 | ||
| BC100753.1 | - | 1 - 2 | 495 | 90 | ||
| BC100754.1 | - | 1 - 2 | 489 | 90 | ||
| BC141283.1 | - | 1 - 2 | 1021 | 9055895 | 90 | |
| L41145.1 | - | 1 - 2 | 490 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmp5
Genomic Map