Assay Details
| Gene Symbol: Lrrc49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146046.1 | NP_001139518.1 | 12 - 13 | 1679 | 107 | |
| NM_001146047.1 | NP_001139519.1 | 13 - 14 | 1678 | 107 | ||
| NM_145616.4 | NP_663591.3 | 13 - 14 | 1877 | 107 | ||
| GenBank mRNA | AK049268.1 | - | 13 - 14 | 1656 | 107 | |
| AK051325.1 | - | 12 - 13 | 1484 | 107 | ||
| AK082406.1 | - | 908 | 107 | |||
| AK085022.1 | - | 1767 | 107 | |||
| BC016574.1 | - | 12 - 13 | 1446 | 3594577 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc49
Genomic Map