Assay Details
| Gene Symbol: Calml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138304.2 | NP_612177.1 | 1 - 2 | 54 | 120 | |
| GenBank mRNA | AY061807.1 | - | 1 - 2 | 54 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY061807.1, NM_138304.2 transcripts.
Set Membership: |
|

Calml4
Genomic Map