Assay Details
| Gene Symbol: Calml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138304.2 | NP_612177.1 | 2 - 3 | 90 | 74 | |
| GenBank mRNA | AK008039.1 | - | 2 - 3 | 76 | 74 | |
| AK146656.1 | - | 2 - 3 | 63 | 74 | ||
| AY061807.1 | - | 2 - 3 | 90 | 74 | ||
| BC013541.1 | - | 2 - 3 | 68 | 4241711 | 74 | |
| BC023475.1 | - | 2 - 3 | 55 | 5042406 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008039.1, AK146656.1, AY061807.1, BC013541.1, BC023475.1, NM_138304.2 transcripts.
Set Membership: |
|

Calml4
Genomic Map