Assay Details
| Gene Symbol: Rexo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024233.3 | NP_077195.2 | 2 - 3 | 286 | 69 | |
| GenBank mRNA | AK007731.1 | - | 2 - 3 | 288 | 69 | |
| AK090202.1 | - | 2 - 3 | 290 | 69 | ||
| AK150287.1 | - | 2 - 3 | 202 | 69 | ||
| AK171619.1 | - | 2 - 3 | 189 | 69 | ||
| AK172641.1 | - | 2 - 3 | 264 | 69 | ||
| BC003445.1 | - | 2 - 3 | 222 | 2651110 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rexo2
Genomic Map