Assay Details
| Gene Symbol: Ireb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022655.3 | NP_073146.2 | 6 - 7 | 826 | 68 | |
| GenBank mRNA | BC044665.1 | - | 6 - 7 | 793 | 2647641 | 68 |
| CK636500.1 | - | 2 - 3 | 118 | 68 | ||
| CN534597.1 | - | 4 - 5 | 480 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ireb2
Genomic Map