Assay Details
| Gene Symbol: Ireb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022655.3 | NP_073146.2 | 9 - 10 | 1312 | 76 | |
| GenBank mRNA | BC044665.1 | - | 9 - 10 | 1279 | 2647641 | 76 |
| CK636500.1 | - | 5 - 6 | 604 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ireb2
Genomic Map