Assay Details
| Gene Symbol: Wdr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025375.2 | NP_001020546.1 | 8 - 9 | 879 | 68 | |
| NM_001025376.2 | NP_001020547.1 | 8 - 9 | 768 | 68 | ||
| NM_023191.3 | NP_075680.1 | 8 - 9 | 773 | 68 | ||
| GenBank mRNA | AF309554.1 | - | 8 - 9 | 731 | 68 | |
| AK010283.1 | - | 8 - 9 | 719 | 68 | ||
| AK012342.1 | - | 8 - 9 | 785 | 68 | ||
| AK013114.1 | - | 8 - 9 | 737 | 68 | ||
| AK077037.1 | - | 813 | 68 | |||
| AK142498.1 | - | 8 - 9 | 736 | 68 | ||
| AK167706.1 | - | 8 - 9 | 806 | 68 | ||
| AK205840.1 | - | 2 - 3 | 165 | 68 | ||
| BC023026.1 | - | 8 - 9 | 718 | 5374890 | 68 | |
| EF011623.1 | - | 8 - 9 | 716 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr61
Genomic Map