Assay Details
| Gene Symbol: Pkib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039050.1 | NP_001034139.1 | 3 - 4 | 587 | 111 | |
| NM_001039051.2 | NP_001034140.1 | 3 - 4 | 393 | 111 | ||
| NM_001039052.1 | NP_001034141.1 | 3 - 4 | 253 | 111 | ||
| NM_001039053.2 | NP_001034142.2 | 4 - 5 | 289 | 111 | ||
| NM_001190402.1 | NP_001177331.1 | 3 - 4 | 491 | 111 | ||
| GenBank mRNA | AK005281.1 | - | 3 - 4 | 325 | 111 | |
| AK156950.1 | - | 3 - 4 | 280 | 111 | ||
| AK157521.1 | - | 3 - 4 | 281 | 111 | ||
| BC052351.1 | - | 3 - 4 | 587 | 111 | ||
| BC061162.1 | - | 3 - 4 | 253 | 111 | ||
| BC087894.1 | - | 3 - 4 | 478 | 6771715 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156950.1, AK157521.1, BC061162.1, BC087894.1, NM_001039050.1, NM_001039051.2, NM_001039052.1, NM_001039053.2, NM_001190402.1 transcripts.
Set Membership: |
|

Pkib
Genomic Map