Assay Details
| Gene Symbol: Smpdl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020561.2 | NP_065586.3 | 4 - 5 | 694 | 76 | |
| GenBank mRNA | AK002455.1 | - | 4 - 5 | 633 | 76 | |
| AK151683.1 | - | 4 - 5 | 580 | 76 | ||
| AK152282.1 | - | 4 - 5 | 595 | 76 | ||
| AK196251.1 | - | 2 - 3 | 166 | 76 | ||
| BC139288.1 | - | 4 - 5 | 586 | 8862310 | 76 | |
| BC139289.1 | - | 4 - 5 | 586 | 8862311 | 76 | |
| Y08135.1 | - | 4 - 5 | 648 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smpdl3a
Genomic Map