Assay Details
| Gene Symbol: Grik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081097.2 | NP_001074566.1 | 11 - 12 | 1753 | 70 | |
| GenBank mRNA | AF245444.1 | - | 3 - 4 | 548 | 70 | |
| BC118004.1 | - | 11 - 12 | 1748 | 70 | ||
| CF744155.1 | - | 2 - 3 | 406 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik3
Genomic Map