Assay Details
| Gene Symbol: Grik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081097.2 | NP_001074566.1 | 7 - 8 | 1107 | 64 | |
| GenBank mRNA | BC118004.1 | - | 7 - 8 | 1102 | 64 | |
| BU611628.1 | - | 2 - 3 | 275 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik3
Genomic Map