Assay Details
| Gene Symbol: Grik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081097.2 | NP_001074566.1 | 8 - 9 | 1213 | 89 | |
| GenBank mRNA | BC118004.1 | - | 8 - 9 | 1208 | 89 | |
| BU611628.1 | - | 3 - 4 | 381 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik3
Genomic Map