Assay Details
| Gene Symbol: Grik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081097.2 | NP_001074566.1 | 9 - 10 | 1325 | 79 | |
| GenBank mRNA | AF245444.1 | - | 1 - 2 | 120 | 79 | |
| BC118004.1 | - | 9 - 10 | 1320 | 79 | ||
| BU611628.1 | - | 4 - 5 | 493 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik3
Genomic Map