Assay Details
| Gene Symbol: Eva1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172145.3 | NP_742157.1 | 1 - 2 | 624 | 99 | |
| GenBank mRNA | AK076006.1 | - | 1 - 2 | 626 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076006.1, NM_172145.3 transcripts.
Set Membership: |
|

Eva1b
Genomic Map