Assay Details
| Gene Symbol: Zmym1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026670.4 | NP_080946.3 | 2 - 3 | 288 | 192 | |
| GenBank mRNA | AK016783.1 | - | 2 - 3 | 292 | 192 | |
| AK035618.1 | - | 2 - 3 | 288 | 192 | ||
| AK044538.1 | - | 2 - 3 | 298 | 192 | ||
| AK049568.1 | - | 2 - 3 | 278 | 192 | ||
| AK138687.1 | - | 2 - 3 | 250 | 192 | ||
| AK166769.1 | - | 2 - 3 | 271 | 192 | ||
| AK167508.1 | - | 2 - 3 | 294 | 192 | ||
| BC027750.1 | - | 2 - 3 | 315 | 192 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049568.1, AK167508.1, BC027750.1, NM_026670.4 transcripts.
Set Membership: |
|

Zmym1
Genomic Map