Assay Details
| Gene Symbol: Smim12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030252.2 | NP_084528.1 | 1 - 2 | 114 | 63 | |
| GenBank mRNA | AK075968.1 | - | 1 - 2 | 89 | 63 | |
| AK131915.1 | - | 1 - 2 | 79 | 63 | ||
| AK141221.1 | - | 1 - 2 | 48 | 63 | ||
| BC147067.1 | - | 1 - 2 | 76 | 9056070 | 63 | |
| BC147068.1 | - | 1 - 2 | 76 | 9056074 | 63 | |
| BY237825.1 | - | 1 - 2 | 115 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK075968.1,AK131915.1,AK141221.1,BC147067.1,BC147068.1,NM_030252.2 transcripts.
This assay is in the 5'UTR region of the AK075968.1,AK131915.1,AK141221.1,BC147067.1,BC147068.1,NM_030252.2 transcripts.
Set Membership: |
|

Smim12
Genomic Map