Assay Details
| Gene Symbol: AU040320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035525.1 | NP_001030602.1 | 3 - 4 | 805 | 69 | |
| NM_001035526.1 | NP_001030603.1 | 3 - 4 | 923 | 69 | ||
| NM_133886.2 | NP_598647.1 | 3 - 4 | 923 | 69 | ||
| GenBank mRNA | AK031342.1 | - | 3 - 4 | 867 | 69 | |
| AK043006.1 | - | 3 - 4 | 873 | 69 | ||
| AK049570.1 | - | 3 - 4 | 806 | 69 | ||
| AK084668.1 | - | 3 - 4 | 768 | 69 | ||
| AK147569.1 | - | 3454 | 69 | |||
| AK170261.1 | - | 3 - 4 | 880 | 69 | ||
| BC028869.1 | - | 3 - 4 | 893 | 4235813 | 69 | |
| CB249000.1 | - | 1 - 2 | 281 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

AU040320
Genomic Map