Assay Details
| Gene Symbol: Gjb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008127.4 | NP_032153.1 | 2 | 1320 | 88 | |
| GenBank mRNA | AK076408.1 | - | 2 | 1320 | 88 | |
| BC064060.1 | - | 1 | 1258 | 1498230 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972381 | BC064060.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972381 | NM_008127.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907904 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420259 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144865 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905650 | AK002285.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905650 | AK004049.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905650 | AK007337.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905650 | AK019922.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905650 | AK137906.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905650 | AK147477.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905650 | AK216319.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905650 | BC024336.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905650 | NM_023260.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759760 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176608 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115506 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972381 | BC064060.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972381 | NM_008127.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P187121 | BC064060.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P187121 | AK076408.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P187121 | NM_008127.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284861 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00983080 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00983084 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115506 | - | - |
| Whole Mouse Genome G4122A | A_51_P187121 | BC064060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187121 | AK076408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187121 | NM_008127.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114325 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115506 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03452084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03452082 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03452083 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03452080 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03452078 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03452073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03300553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03748839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03748828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03748834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03300541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03416672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23284830 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10516488 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516489 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422179_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10516487 | - | - |
| Mouse Genome 430 2.0 Array | 1422179_at | - | - |
| Mouse Genome 430 2.0 Array | 1422179_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14621_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14621_PM_at | - | - |
| Mouse Expression Set 430 | 1422179_PM_at | - | - |
| Mouse Expression Set 430 | 1422179_at | - | - |
| Murine Genome U74v2 Set | 99373_PM_at | - | - |
| Murine Genome U74v2 Set | 99373_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422179_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422179_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422179_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422179_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 582409 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4850133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600190 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600190 | - | - |
| mouse-6 v1.1 (Illumina) | scl0014621.2_189-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0014621.2_189-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600190 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600190 | - | - |

Gjb4
Genomic Map