Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400011264.mm.1 |
NM_023603.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011264.mm.1 |
NM_023603.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011264.mm.1 |
NR_045010.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011264.mm.1 |
NR_045010.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011264.mm.1 |
TC0400001477.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011265.mm.1 |
NM_023603.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011265.mm.1 |
NM_023603.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011265.mm.1 |
NR_045010.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011265.mm.1 |
NR_045010.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011265.mm.1 |
TC0400001477.mm.1 |
MTA TC |