Assay Details
| Gene Symbol: Mcm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027830.2 | NP_082106.2 | 12 - 13 | 2282 | 77 | |
| GenBank mRNA | AK133212.1 | - | 5 - 6 | 584 | 77 | |
| AK133345.1 | - | 5 - 6 | 1397 | 77 | ||
| BC062185.1 | - | 3 - 4 | 359 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm9
Genomic Map