Assay Details
| Gene Symbol: Tube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028006.2 | NP_082282.1 | 10 - 11 | 1161 | 103 | |
| GenBank mRNA | AK010005.1 | - | 10 - 11 | 1155 | 103 | |
| AK040492.1 | - | 2 - 3 | 2719 | 103 | ||
| AK041416.1 | - | 10 - 11 | 1127 | 103 | ||
| AK041986.1 | - | 567 | 103 | |||
| AK087403.1 | - | 9 - 10 | 940 | 103 | ||
| AK148039.1 | - | 10 - 11 | 1161 | 103 | ||
| BC038654.1 | - | 7 - 8 | 1126 | 103 | ||
| BC062179.1 | - | 10 - 11 | 1158 | 6333817 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tube1
Genomic Map