Assay Details
| Gene Symbol: Tube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028006.2 | NP_082282.1 | 1 - 2 | 89 | 69 | |
| GenBank mRNA | AK010005.1 | - | 1 - 2 | 83 | 69 | |
| AK039886.1 | - | 1 - 2 | 86 | 69 | ||
| AK041416.1 | - | 1 - 2 | 54 | 69 | ||
| AK087403.1 | - | 1 - 2 | 44 | 69 | ||
| AK148039.1 | - | 1 - 2 | 89 | 69 | ||
| BC062179.1 | - | 1 - 2 | 86 | 6333817 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010005.1, AK148039.1, BC062179.1, NM_028006.2 transcripts.
Set Membership: |
|

Tube1
Genomic Map