Assay Details
| Gene Symbol: Tube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028006.2 | NP_082282.1 | 8 - 9 | 874 | 113 | |
| GenBank mRNA | AK010005.1 | - | 8 - 9 | 868 | 113 | |
| AK041416.1 | - | 8 - 9 | 839 | 113 | ||
| AK148039.1 | - | 8 - 9 | 874 | 113 | ||
| BC038654.1 | - | 5 - 6 | 839 | 113 | ||
| BC062179.1 | - | 8 - 9 | 871 | 6333817 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tube1
Genomic Map