Assay Details
| Gene Symbol: Tube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028006.2 | NP_082282.1 | 9 - 10 | 1014 | 85 | |
| GenBank mRNA | AK010005.1 | - | 9 - 10 | 1008 | 85 | |
| AK040492.1 | - | 1 - 2 | 2572 | 85 | ||
| AK041416.1 | - | 9 - 10 | 86 | |||
| AK041986.1 | - | 420 | 85 | |||
| AK087403.1 | - | 8 - 9 | 793 | 85 | ||
| AK148039.1 | - | 9 - 10 | 1014 | 85 | ||
| BC038654.1 | - | 6 - 7 | 979 | 85 | ||
| BC062179.1 | - | 9 - 10 | 1011 | 6333817 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tube1
Genomic Map