Assay Details
| Gene Symbol: Frk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159544.1 | NP_001153016.1 | 1 - 2 | 970 | 86 | |
| NM_010237.3 | NP_034367.2 | 2 - 3 | 814 | 86 | ||
| GenBank mRNA | AK028855.1 | - | 2 - 3 | 754 | 86 | |
| AK052614.1 | - | 2 - 3 | 686 | 86 | ||
| AK077264.1 | - | 1 - 2 | 839 | 86 | ||
| AK085191.1 | - | 1 - 2 | 815 | 86 | ||
| AK166379.1 | - | 2 - 3 | 814 | 86 | ||
| BC007137.1 | - | 2 - 3 | 827 | 3601246 | 86 | |
| L36132.1 | - | 1 - 2 | 849 | 86 | ||
| Z48757.1 | - | 2 - 3 | 679 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frk
Genomic Map