Assay Details
| Gene Symbol: Frk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159544.1 | NP_001153016.1 | 4 - 5 | 1425 | 87 | |
| NM_010237.3 | NP_034367.2 | 5 - 6 | 1269 | 87 | ||
| GenBank mRNA | AK028855.1 | - | 5 - 6 | 1209 | 87 | |
| AK052614.1 | - | 5 - 6 | 1141 | 87 | ||
| AK077264.1 | - | 4 - 5 | 1294 | 87 | ||
| AK085191.1 | - | 4 - 5 | 1270 | 87 | ||
| AK166379.1 | - | 5 - 6 | 1269 | 87 | ||
| BC007137.1 | - | 5 - 6 | 1282 | 3601246 | 87 | |
| L36132.1 | - | 4 - 5 | 1304 | 87 | ||
| Z48757.1 | - | 5 - 6 | 1134 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frk
Genomic Map