Assay Details
| Gene Symbol: Ogdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252282.1 | NP_001239211.1 | 9 - 10 | 1196 | 84 | |
| NM_001252283.1 | NP_001239212.1 | 9 - 10 | 1184 | 84 | ||
| NM_001252287.1 | NP_001239216.1 | 8 - 9 | 1227 | 84 | ||
| NM_001252288.1 | NP_001239217.1 | 8 - 9 | 1199 | 84 | ||
| NM_010956.4 | NP_035086.2 | 8 - 9 | 1151 | 84 | ||
| GenBank mRNA | AK147289.1 | - | 8 - 9 | 1152 | 84 | |
| AK150009.1 | - | 8 - 9 | 1117 | 84 | ||
| AK169286.1 | - | 8 - 9 | 1143 | 84 | ||
| AK188421.1 | - | 1 - 2 | 81 | 84 | ||
| AK189909.1 | - | 185 | 84 | |||
| AK220536.1 | - | 9 - 10 | 1158 | 84 | ||
| BC013670.1 | - | 8 - 9 | 1085 | 4188988 | 84 | |
| BC049104.1 | - | 8 - 9 | 1152 | 6417392 | 84 | |
| BC057354.1 | - | 9 - 10 | 1112 | 6827509 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogdh
Genomic Map