Assay Details
| Gene Symbol: Clic4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013885.2 | NP_038913.1 | 2 - 3 | 355 | 108 | |
| GenBank mRNA | AF102578.1 | - | 2 - 3 | 353 | 108 | |
| AK031211.1 | - | 2 - 3 | 362 | 108 | ||
| AK049191.1 | - | 2 - 3 | 360 | 108 | ||
| AK049255.1 | - | 2 - 3 | 360 | 108 | ||
| AK054024.1 | - | 2 - 3 | 360 | 108 | ||
| AK150993.1 | - | 2 - 3 | 359 | 108 | ||
| AK152273.1 | - | 2 - 3 | 361 | 108 | ||
| AK153432.1 | - | 2 - 3 | 362 | 108 | ||
| AK155826.1 | - | 2 - 3 | 359 | 108 | ||
| AK159402.1 | - | 2 - 3 | 362 | 108 | ||
| AK160856.1 | - | 2 - 3 | 361 | 108 | ||
| AK161034.1 | - | 2 - 3 | 361 | 108 | ||
| AK166244.1 | - | 2 - 3 | 362 | 108 | ||
| AK166794.1 | - | 2 - 3 | 359 | 108 | ||
| AK169376.1 | - | 2 - 3 | 359 | 108 | ||
| AK169834.1 | - | 2 - 3 | 359 | 108 | ||
| AK170942.1 | - | 2 - 3 | 410 | 108 | ||
| AK171243.1 | - | 2 - 3 | 362 | 108 | ||
| AK172026.1 | - | 2 - 3 | 361 | 108 | ||
| AK172516.1 | - | 2 - 3 | 335 | 108 | ||
| AK172660.1 | - | 2 - 3 | 359 | 108 | ||
| AK211909.1 | - | 1 - 2 | 62 | 108 | ||
| BC046384.1 | - | 2 - 3 | 339 | 3155573 | 108 | |
| BC052890.1 | - | 2 - 3 | 343 | 30056606 | 108 | |
| CT010279.1 | - | 2 - 3 | 184 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clic4
Genomic Map