Assay Details
| Gene Symbol: Ipcef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033391.2 | NP_001028563.1 | 4 - 5 | 235 | 74 | |
| NM_001170800.1 | NP_001164271.1 | 4 - 5 | 533 | 74 | ||
| NM_001170801.1 | NP_001164272.1 | 4 - 5 | 533 | 74 | ||
| NM_001170802.1 | NP_001164273.1 | 4 - 5 | 533 | 74 | ||
| GenBank mRNA | AK081159.1 | - | 4 - 5 | 383 | 74 | |
| AK135851.1 | - | 261 | 74 | |||
| AK157384.1 | - | 4 - 5 | 534 | 74 | ||
| AK220339.1 | - | 3 - 4 | 342 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ipcef1
Genomic Map