Assay Details
| Gene Symbol: Zmym6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285885.1 | NP_001272814.1 | 4 - 5 | 1628 | 77 | |
| NM_001285887.1 | NP_001272816.1 | 5 - 6 | 821 | 77 | ||
| NM_001285888.1 | NP_001272817.1 | 5 - 6 | 828 | 77 | ||
| NM_177462.5 | NP_803413.1 | 5 - 6 | 821 | 77 | ||
| NR_104366.1 | 5 - 6 | 1884 | 77 | |||
| GenBank mRNA | AK035136.1 | - | 5 - 6 | 823 | 77 | |
| AK081836.1 | - | 5 - 6 | 811 | 77 | ||
| AK082594.1 | - | 4 - 5 | 1352 | 77 | ||
| AK170695.1 | - | 5 - 6 | 1844 | 77 | ||
| BC034119.1 | - | 3 - 4 | 473 | 77 | ||
| BC040744.1 | - | 3 - 4 | 473 | 77 | ||
| BC094598.1 | - | 4 - 5 | 1601 | 77 | ||
| BC140958.1 | - | 5 - 6 | 696 | 9055570 | 77 | |
| BC144807.1 | - | 5 - 6 | 703 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zmym6
Genomic Map