Assay Details
| Gene Symbol: Moxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021509.5 | NP_067484.2 | 4 - 5 | 749 | 115 | |
| GenBank mRNA | AB041606.1 | - | 4 - 5 | 684 | 115 | |
| AK014340.1 | - | 4 - 5 | 750 | 115 | ||
| AK030274.1 | - | 4 - 5 | 690 | 115 | ||
| AK081586.1 | - | 4 - 5 | 715 | 115 | ||
| AK162030.1 | - | 4 - 5 | 691 | 115 | ||
| BC025892.1 | - | 4 - 5 | 447 | 115 | ||
| BC037684.1 | - | 4 - 5 | 687 | 115 | ||
| BC057652.1 | - | 4 - 5 | 663 | 4017199 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Moxd1
Genomic Map