Assay Details
| Gene Symbol: Moxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021509.5 | NP_067484.2 | 5 - 6 | 924 | 75 | |
| GenBank mRNA | AB041606.1 | - | 5 - 6 | 859 | 75 | |
| AK014340.1 | - | 5 - 6 | 925 | 75 | ||
| AK030274.1 | - | 5 - 6 | 865 | 75 | ||
| AK081586.1 | - | 5 - 6 | 890 | 75 | ||
| AK162030.1 | - | 5 - 6 | 866 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Moxd1
Genomic Map