Assay Details
| Gene Symbol: Moxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021509.5 | NP_067484.2 | 7 - 8 | 1199 | 87 | |
| GenBank mRNA | AB041606.1 | - | 7 - 8 | 1134 | 87 | |
| AK014340.1 | - | 7 - 8 | 1200 | 87 | ||
| AK030274.1 | - | 7 - 8 | 1140 | 87 | ||
| AK081586.1 | - | 7 - 8 | 1165 | 87 | ||
| AK162030.1 | - | 7 - 8 | 1141 | 87 | ||
| BC025892.1 | - | 7 - 8 | 897 | 87 | ||
| BC037684.1 | - | 7 - 8 | 1137 | 87 | ||
| BC057652.1 | - | 7 - 8 | 1113 | 4017199 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Moxd1
Genomic Map