Assay Details
| Gene Symbol: Moxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021509.5 | NP_067484.2 | 8 - 9 | 1384 | 107 | |
| GenBank mRNA | AB041606.1 | - | 8 - 9 | 1319 | 107 | |
| AK014340.1 | - | 8 - 9 | 1385 | 107 | ||
| AK030274.1 | - | 8 - 9 | 1325 | 107 | ||
| AK081586.1 | - | 8 - 9 | 1350 | 107 | ||
| AK162030.1 | - | 8 - 9 | 1326 | 107 | ||
| BC025892.1 | - | 8 - 9 | 1082 | 107 | ||
| BC037684.1 | - | 8 - 9 | 1322 | 107 | ||
| BC057652.1 | - | 8 - 9 | 1298 | 4017199 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Moxd1
Genomic Map