Assay Details
| Gene Symbol: Rsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023665.3 | NP_076154.3 | 4 - 5 | 972 | 118 | |
| GenBank mRNA | AK012363.1 | - | 4 - 5 | 973 | 118 | |
| AK150024.1 | - | 4 - 5 | 973 | 118 | ||
| AK150665.1 | - | 4 - 5 | 952 | 118 | ||
| AK152736.1 | - | 4 - 5 | 952 | 118 | ||
| AK178549.1 | - | 3 - 4 | 318 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325561 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325563 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325564 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131766 | BC054728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131766 | BC056986.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131766 | BC046231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131766 | BC043057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131766 | AK169760.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72611 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131766 | BC054728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131766 | BC056986.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131766 | BC046231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131766 | BC043057.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131766 | AK169760.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23325579 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00995655 | - | - |
| Whole Mouse Genome G4122A | A_51_P346715 | BC054728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346715 | BC056986.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346715 | BC046231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346715 | BC043057.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346715 | AK169760.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113539 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00995655 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03778611 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325563 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23325561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23325560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03221574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03327312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03986317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03610285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03221573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23325567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03778605 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10509019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509022 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509021 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417001_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435357_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448538_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10509014 | - | - |
| Mouse Genome 430 2.0 Array | 1435357_at | - | - |
| Mouse Genome 430 2.0 Array | 1417001_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448538_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435357_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417001_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448538_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27981_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27981_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400013130.mm.1 | NM_023665.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013130.mm.1 | NM_023665.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400013130.mm.1 | TC0400001646.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417001_PM_a_at | - | - |
| Mouse Expression Set 430 | 1435357_at | - | - |
| Mouse Expression Set 430 | 1435357_PM_at | - | - |
| Mouse Expression Set 430 | 1417001_a_at | - | - |
| Mouse Expression Set 430 | 1448538_a_at | - | - |
| Mouse Expression Set 430 | 1448538_PM_a_at | - | - |
| Murine Genome U74v2 Set | 92542_PM_at | - | - |
| Murine Genome U74v2 Set | 92542_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448538_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417001_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435357_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417001_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448538_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435357_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448538_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435357_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448538_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435357_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417001_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417001_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6520114 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520114 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2731439 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2731439 | - | - |
| mouse-6 v1.1 (Illumina) | scl24840.4_10-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520114 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24840.4_10-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520114 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2731439 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2731439 | - | - |

Rsrp1
Genomic Map