Assay Details
| Gene Symbol: Srrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130477.1 | NP_001123949.1 | 4 - 5 | 499 | 131 | |
| NM_016799.3 | NP_058079.2 | 4 - 5 | 499 | 131 | ||
| GenBank mRNA | AF062655.1 | - | 4 - 5 | 425 | 131 | |
| AK008284.1 | - | 4 - 5 | 427 | 131 | ||
| AK028412.1 | - | 4 - 5 | 431 | 131 | ||
| BC079542.1 | - | 3 - 4 | 2484 | 131 | ||
| BC094322.1 | - | 4 - 5 | 414 | 6401250 | 131 | |
| BU514596.1 | - | 4 - 5 | 499 | 131 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Srrm1
Genomic Map