Assay Details
| Gene Symbol: P3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042411.1 | NP_001035874.1 | 12 - 13 | 1526 | 85 | |
| NM_001286148.1 | NP_001273077.1 | 12 - 13 | 1913 | 85 | ||
| NM_019782.3 | NP_062756.2 | 12 - 13 | 1892 | 85 | ||
| NM_019783.2 | NP_062757.2 | 12 - 13 | 1892 | 85 | ||
| GenBank mRNA | AK010578.1 | - | 12 - 13 | 1894 | 85 | |
| AK030436.1 | - | 12 - 13 | 1890 | 85 | ||
| AK132262.1 | - | 12 - 13 | 1889 | 85 | ||
| AK159505.1 | - | 12 - 13 | 1527 | 85 | ||
| BC024047.1 | - | 12 - 13 | 1858 | 5148677 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

P3h1
Genomic Map