Assay Details
| Gene Symbol: Gm6150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038036.1 | 3 - 4 | 489 | 155 | ||
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0100000042.rn
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84517 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133421 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06972291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08815902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08252136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08367792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08515409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08525103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08252137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08868752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08491481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26931739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08581133 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1439338_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439338_at | - | - |
| Mouse Genome 430 2.0 Array | 1439338_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000000479.mm.1 | NR_038036.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000000479.mm.1 | NR_038036.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000000479.mm.1 | TC1000000067.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439338_at | - | - |
| Mouse Expression Set 430 | 1439338_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 595375 | - | - |

Gm6150
Genomic Map