| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000012582.mm.1 |
NM_001081344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012582.mm.1 |
NM_001081344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012582.mm.1 |
TC1000001737.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012595.mm.1 |
NM_001081344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012595.mm.1 |
NM_001081344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012595.mm.1 |
TC1000001737.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012584.mm.1 |
NM_001081344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012584.mm.1 |
NM_001081344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012584.mm.1 |
TC1000001737.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012597.mm.1 |
NM_001081344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012597.mm.1 |
NM_001081344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012597.mm.1 |
TC1000001737.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012591.mm.1 |
NM_001081344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012591.mm.1 |
NM_001081344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012591.mm.1 |
TC1000001737.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012594.mm.1 |
NM_001081344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012594.mm.1 |
NM_001081344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012594.mm.1 |
TC1000001737.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012583.mm.1 |
NM_001081344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012583.mm.1 |
NM_001081344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012583.mm.1 |
TC1000001737.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012596.mm.1 |
NM_001081344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012596.mm.1 |
NM_001081344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012596.mm.1 |
TC1000001737.mm.1 |
MTA TC |