Assay Details
| Gene Symbol: Hey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013904.1 | NP_038932.1 | 3 - 4 | 386 | 92 | |
| GenBank mRNA | AB093589.1 | - | 3 - 4 | 254 | 92 | |
| AF172287.1 | - | 3 - 4 | 386 | 92 | ||
| AF173902.1 | - | 3 - 4 | 378 | 92 | ||
| AF232240.1 | - | 3 - 4 | 382 | 92 | ||
| AJ271867.1 | - | 3 - 4 | 386 | 92 | ||
| AK031506.1 | - | 3 - 4 | 233 | 92 | ||
| AK158000.1 | - | 3 - 4 | 387 | 92 | ||
| BC103575.1 | - | 3 - 4 | 326 | 40045790 | 92 | |
| BC103576.1 | - | 3 - 4 | 329 | 40045791 | 92 | |
| BC105651.1 | - | 3 - 4 | 326 | 40046419 | 92 | |
| BC105652.1 | - | 3 - 4 | 329 | 40046420 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hey2
Genomic Map